Learn More
Abcam Sitravatinib, 25MG
Catalog No. 502681670
Encompass_Preferred
MW 629.7 Da, Purity =98%. Broad spectrum receptor tyrosine kinase inhibitor (e.g. Axl, c-Met, PDGFR, VEGFR, Ephrin receptor family, and FLT3). Better efficacy in mouse models than imatinib and crizotinib.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-1670 Supplier Abcam Supplier No. AB28242225MG
MW 629.7 Da, Purity =98%. Broad spectrum receptor tyrosine kinase inhibitor (e.g. Axl, c-Met, PDGFR, VEGFR, Ephrin receptor family, and FLT3). Better efficacy in mouse models than imatinib and crizotinib.
The product is subject to the following: Abcam Restricted Use Statement