Learn More
Selleck Chemical LLC Sitravatinib (MGCD516) 5mg 1123837-84-2 MG-516
SDP
Supplier:  Selleck Chemical LLC S85735MG
Encompass_Preferred
Sitravatinib (MGCD516, MG-516) is a novel small molecule inhibitor targeting multiple RTKs involved in driving sarcoma cell growth, including c-Kit, PDGFR?, PDGFR?, c-Met, and Axl. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8108