Learn More
Electron Microscopy Sciences 4-Acetamido-4'-Isothiocyanostilbene-2,2'-Disulfonic Acid 250 MG
SDP
Supplier:  Electron Microscopy Sciences 10030
Encompass_Preferred
CAS #51023-76-8 C₁₇H₁₂N₂O7S₃Na₂ F.W. 498.50
SITS has been used as a vital stain, as a label for immunostaining, and as a neuronal tracer. SITS is also a non-penetrating fluorescent marker for erythrocyte membranes which acts as a potent inhibitor of anion transport. For more information related to the preparation and characterization of SITS, please refer to: Jakobsen, P. and Horobin, Richard W.(1989). Stain Technology, Vol.64, No.6 pg.301-313. 4-Acetamido-4'-Isothiocyanostilbene-2,2'-Disulfonic Acid.
Catalog No. 50-980-150