Learn More
REPLIGEN CORPORATION Repligen TangenX SIUS TFF Cassette, 2.5 m² Membrane Area, Single-use, 100 kDa Pore Size
SDP
Supplier:  REPLIGEN CORPORATION XP100G25L
Encompass
The Repligen TangenX SIUS TFF Cassette is a single-use filtration module designed for efficient downstream bioprocessing. With a membrane surface area of 2.5 m², it streamlines ultrafiltration and diafiltration processes by reducing labor, buffer, and water usage through its pre-sanitized design. Its high flux capabilities and compatibility with existing equipment differentiate it from other products in the market, empowering biopharmaceutical purification with scalability and improved yield.
Catalog No. NC1804761