Learn More
REPLIGEN CORPORATION Repligen TangenX SIUS Manual Clamp, 4-Bolt Configuration, Isolated Process Fluid, 52 lbs
SDP
Supplier:  REPLIGEN CORPORATION TSPDI4BMC
Encompass
The Repligen TangenX™ SIUS™ Horizontal Cassette Clamp is a 4-bolt manual clamp designed for tangential flow filtration (TFF) applications. It features a robust stainless steel construction that isolates process fluid, ensuring purity and preventing contamination. With compatibility for industry-standard cassettes and a maximum flow rate of 49 LPM, this clamp provides reliable performance and a liquid-tight seal for optimal filtration efficiency.
Catalog No. NC0706586
Add to Cart
Request a Quote