Learn More
REPLIGEN CORPORATION TangenX SIUS TFF Cassette, 100 kD MWCO, 0.5 m² Surface Area, Single-Use
SDP
Supplier:  REPLIGEN CORPORATION XP100G05L
Encompass
The Repligen TangenX SIUS TFF Cassette is a single-use filtration solution, designed for efficient ultrafiltration and diafiltration in scientific and bioprocessing applications. Its optimized design reduces the need for extensive cleaning processes, saving labor and resources. With a 100 kD molecular weight cutoff and 0.5 m² surface area, it delivers consistent hydraulic performance from development through to large-scale manufacturing, distinguishing it from traditional reusable systems.
Catalog No. NC1095082