Learn More
ADVANCED BIOSCIENCE LABORATORIES INC SIV p27 Antigen Capture Assay, 96-well microplate, double antibody sandwich enzyme immunoassay, colorimetric ELISA, target analyte SIV p27 antigen
Supplier:  ADVANCED BIOSCIENCE LABORATORIES INC 5436
Encompass
SIV p27 Antigen Capture Assay, 96-well microplate, double antibody sandwich enzyme immunoassay, colorimetric ELISA, target analyte SIV p27 antigen
Catalog No. NC1445248