Learn More
Abcam Sivelestat, neutrophil elastase inhibitor, 25MG
Catalog No. 502704870
Encompass_Preferred
MW 434.5 Da. Potent, selective, substrate competitive neutrophil elastase inhibitor (IC₅₀ values are 19-49 nM and 5.6 μM for leukocyte and pancreas elastase respectively). Suppresses neutrophil-induced capillary permeability and leukocyte kinetics. Active in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4870 Supplier Abcam Supplier No. AB14618425MG
MW 434.5 Da. Potent, selective, substrate competitive neutrophil elastase inhibitor (IC₅₀ values are 19-49 nM and 5.6 μM for leukocyte and pancreas elastase respectively). Suppresses neutrophil-induced capillary permeability and leukocyte kinetics. Active in vivo. .
The product is subject to the following: Abcam Restricted Use Statement