Learn More
Selleck Chemical LLC Sivelestat (ONO-5046) 10mg 127373-66-4 ONO5046, LY544349, EI546
SDP
Catalog No. 501365701
Encompass_Preferred
Sivelestat (ONO5046, LY544349, EI546) is a potent and selective inhibitor of neutrophil elastase with IC50 of 44nM. It almost shows no activity at a range of other proteases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5701 Supplier Selleck Chemical LLC Supplier No. S813610MG
Sivelestat (ONO5046, LY544349, EI546) is a potent and selective inhibitor of neutrophil elastase with IC50 of 44nM. It almost shows no activity at a range of other proteases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.