Learn More
Medchemexpress LLC Sivelestat sodium tetrahydrate | 201677-61-4 | 99.9% | 528.51 | 1 ML
SDP
Catalog No. 5000386250
Encompass_Preferred
Sivelestat sodium tetrahydrate is a competitive inhibitor of human neutrophil elastase, with an IC50 of 44 nM and a Ki of 200 nM. It has the potential for the study of acute lung injury/acute respiratory distress syndrome or disseminated intravascular coagulation in COVID-19.
  • Competitively inhibits human neutrophil elastase.
  • Does not inhibit other proteases like trypsin, thrombin, plasmin, plasma kallikrein, pancreas kallikrein, chymotrypsin, and cathepsin G.
  • Suppresses lung hemorrhage.
  • Ameliorates lung injury after hemorrhagic shock.
  • Prevents ischemia-reperfusion injury in the bladder.
Catalog No. 50-003-86250 Supplier Medchemexpress LLC Supplier No. HY17443B10MM/1ML
Sivelestat sodium tetrahydrate is a competitive inhibitor of human neutrophil elastase, with an IC50 of 44 nM and a Ki of 200 nM. It has the potential for the study of acute lung injury/acute respiratory distress syndrome or disseminated intravascular coagulation in COVID-19.
  • Competitively inhibits human neutrophil elastase.
  • Does not inhibit other proteases like trypsin, thrombin, plasmin, plasma kallikrein, pancreas kallikrein, chymotrypsin, and cathepsin G.
  • Suppresses lung hemorrhage.
  • Ameliorates lung injury after hemorrhagic shock.
  • Prevents ischemia-reperfusion injury in the bladder.