Learn More
Cayman Chemical Sivelestat (sodium salt hydrate), Purity: ≥98%, CAS: 201677-61-4, 25 mg
SDP
Catalog No. 502967564
Encompass_Preferred
An inhibitor of neutrophil elastase (IC50 = 44 nM for the human enzyme); selective for neutrophil elastase over trypsin, thrombin, plasmin, kallikrein, chymotrypsin, and cathepsin G at 100 µM; decreases LPS-induced NF-κB nuclear translocation in isolated rat PMVECs at 50 mM; reduces pulmonary edema, increases in the number of MPO-positive cells in the lung tissue, and the severity of lung injury in a rat model of LPS-induced acute lung injury at 10 mg/kg
Catalog No. 50-296-7564 Supplier Cayman Chemical Supplier No. 1777925 MG
An inhibitor of neutrophil elastase (IC50 = 44 nM for the human enzyme); selective for neutrophil elastase over trypsin, thrombin, plasmin, kallikrein, chymotrypsin, and cathepsin G at 100 µM; decreases LPS-induced NF-κB nuclear translocation in isolated rat PMVECs at 50 mM; reduces pulmonary edema, increases in the number of MPO-positive cells in the lung tissue, and the severity of lung injury in a rat model of LPS-induced acute lung injury at 10 mg/kg