Learn More
Thermo Scientific SIX CATION STANDARD II 50ML
Supplier:  Thermo Scientific 046070
Encompass
Increase your productivity and reduce standard preparation errors with Thermo Scientific™ Dionex™ Ion Standards for ion chromatography. Available for routine anion and cation determinations, these single and combined ion standards are made with high purity, reagent-grade chemicals and water. Each standard is NIST traceable and includes a Certificate of Analysis.
Catalog No. NC1145543