Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Six-Channel Syringe Pump; 115 VAC
Catalog No. NC1684242
Encompass
Programming commands allow dispensing of a specified amount, automatic rates changes, direction changes, programmed pauses, external or user input, and sending or receive signals from other equipment. Non-volatile memory restores all setup parameters and
Catalog No. NC1684242 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7590050
Programming commands allow dispensing of a specified amount, automatic rates changes, direction changes, programmed pauses, external or user input, and sending or receive signals from other equipment. Non-volatile memory restores all setup parameters and