Learn More
Waters Corp Waters Six-Column Select Valve Kit, Automated, 14-Port, 6-Position, HPLC
Catalog No. 50791630 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Six-Column Select Valve Kit is an automated 14-port, 6-position switching valve. It is designed for Alliance HPLC series Column Heater and Column Heater/Cooler modules, enabling automated column and method switching for up to six HPLC columns.
  • Provides automated switching capacity for up to six HPLC columns.
  • Allows for column/method switching.
  • Enables sequential running of multiple methods.
  • Supports unattended methods development.
  • Includes a fourteen-port, six-position column selection valve, pre-installed on a mounting plate.
  • Mounting plate slides easily onto pins inside Column Heater or Column Heater/Cooler Module valve compartment.
  • Includes tubing and fittings for connecting the valve to columns or other external devices.
  • Programmable from instrument™s front panel or remotely via Empower™ or MassLynx™ software.
Catalog No. 50-791-630 Supplier Waters Corp Supplier No. 205000164
Add to Cart
Add to Cart
Request a Quote
The Six-Column Select Valve Kit is an automated 14-port, 6-position switching valve. It is designed for Alliance HPLC series Column Heater and Column Heater/Cooler modules, enabling automated column and method switching for up to six HPLC columns.
  • Provides automated switching capacity for up to six HPLC columns.
  • Allows for column/method switching.
  • Enables sequential running of multiple methods.
  • Supports unattended methods development.
  • Includes a fourteen-port, six-position column selection valve, pre-installed on a mounting plate.
  • Mounting plate slides easily onto pins inside Column Heater or Column Heater/Cooler Module valve compartment.
  • Includes tubing and fittings for connecting the valve to columns or other external devices.
  • Programmable from instrument™s front panel or remotely via Empower™ or MassLynx™ software.