Learn More
Aviva Systems Biology Corp Six1 Antibody - N-terminal region (ARP38717_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38717_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Six1. It was validated on Western Blot. . Alternate names: BB138287ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-284-755