Learn More
Aviva Systems Biology Corp SIX1 Antibody - middle region (ARP32375_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32375_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SIX1. Alternate names: BOS3, DFNA23, TIP39ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-54552