Learn More
Aviva Systems Biology Corp Six3 Antibody - C-terminal region (ARP37135_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37135_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Six3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of mouse Six3. Alternate names: Six3a, Six3alpha, Six3b, Six3beta, E130112M24RikApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-284-587