Learn More
Aviva Systems Biology Corp SIX4 Antibody - N-terminal region (ARP39278_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39278_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIX4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SIX4. Alternate names: AREC3, MGC119450, MGC119452, MGC119453ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-56969