Learn More
Medchemexpress LLC Siynfekl Tfa | 99.9% | 1013.14 | 1 MG
SDP
Catalog No. 5000424765
Encompass_Preferred
SIYNFEKL TFA is a variant of the major MHC class I-restricted epitope SIINFEKL. It is an antigenic peptide that can stimulate specific T cells in experimental settings to study competitive interactions between T cells. SIYNFEKL TFA exhibits low affinity for the OT-I T cell receptor (TCR) and can be used for the detection of CD8+ T cells.
  • Can stimulate specific T cells
  • Used to study competitive interactions between T cells
  • Exhibits low affinity for the OT-I T cell receptor (TCR)
  • Can be used for the detection of CD8+ T cells
Catalog No. 50-004-24765 Supplier Medchemexpress LLC Supplier No. HYP10582A1MG
SIYNFEKL TFA is a variant of the major MHC class I-restricted epitope SIINFEKL. It is an antigenic peptide that can stimulate specific T cells in experimental settings to study competitive interactions between T cells. SIYNFEKL TFA exhibits low affinity for the OT-I T cell receptor (TCR) and can be used for the detection of CD8+ T cells.
  • Can stimulate specific T cells
  • Used to study competitive interactions between T cells
  • Exhibits low affinity for the OT-I T cell receptor (TCR)
  • Can be used for the detection of CD8+ T cells