Learn More
Cole-Parmer Cole-Parmer Solid Black Rubber Stoppers, Standard Size 16; 1/Pk
Catalog No. NC1455167 Shop All Cole-Parmer Products
Encompass_Preferred
Black rubber stoppers stay pliant over a long period and withstand laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Contain some sulfur. Autoclavable.
Catalog No. NC1455167 Supplier Cole-Parmer Supplier No. 6299052
Black rubber stoppers stay pliant over a long period and withstand laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Contain some sulfur. Autoclavable.