Learn More
Fine Science Tools SIZE 3 SUTURE THREAD
SDP
Catalog No. NC9321324
Encompass
Product: Suture Thread - Braided Silk/Size 3/0/ Size: 3/0/ Suture Diameter: 0.22mm/ Suture Length: 23m/ Suture Material: Braided Silk/ Sterile: No/ Unit: Each
Catalog No. NC9321324 Supplier Fine Science Tools Supplier No. 1802030
Product: Suture Thread - Braided Silk/Size 3/0/ Size: 3/0/ Suture Diameter: 0.22mm/ Suture Length: 23m/ Suture Material: Braided Silk/ Sterile: No/ Unit: Each