Learn More
STEMCELL Technologies 2 mL Extracellular Vesicle SEC Columns, Size: 2 mL
SDP
Supplier:  STEMCELL Technologies 1000415
Encompass
Extracellular Vesicle Size Exclusion Chromatography (SEC) Columns are ideal for the isolation and purification of extracellular vesicles (EVs) from numerous types of biological matrices, including plasma, serum, and cell culture media, and provide an efficient method for separation of EVs from circulating proteins. Compared to other methods of EV isolation, SEC columns cause minimal alteration in vesicle characteristics such as functionality and shape. The isolated EVs are suitable for downstream analysis including flow cytometry, western blot, nucleic acid extraction, and/or functional assays.The EV isolation protocol using the SEC columns is easy and takes approximately 15 minutes. The columns are available in three sizes for different sample volume ranges and can be used up to five times.
Catalog No. 50-206-9322