Learn More
Chemglass Life Sciences O-Ring, Viton, #107, 12/pk
SDP
Catalog No. NC1876524
Encompass_Preferred
Viton o-ring is stocked in brown 75 durometer. They are suitable for acids, aliphatic hydrocarbons, oxidizing agents and all petroleum products. Viton has a low compression set and high temperature resistance. Viton is especially good for vacuum service and low gas permeability and has a useful temperature range of -30 to 200º C.
Catalog No. NC1876524 Supplier Chemglass Life Sciences Supplier No. CG305107ORING
Viton o-ring is stocked in brown 75 durometer. They are suitable for acids, aliphatic hydrocarbons, oxidizing agents and all petroleum products. Viton has a low compression set and high temperature resistance. Viton is especially good for vacuum service and low gas permeability and has a useful temperature range of -30 to 200º C.