Learn More
Chemglass Life Sciences Stopper, Flask Length, PTFE, Size 27
SDP
Catalog No. CG302006
Encompass_Preferred
Stopper, Flask Length, PTFE, Size 27. PTFE stopper for use in flask length outer joints.
Catalog No. CG302006 Supplier Chemglass Life Sciences Supplier No. CG302006
Stopper, Flask Length, PTFE, Size 27. PTFE stopper for use in flask length outer joints.