Learn More
REFEYN INC SizeFerence Calibrant, tailored for the KaritroMP
SDP
Catalog No. NC3737117
Encompass
SizeFerence is a calibrant tailored for the KaritroMP. This contains a mix of silica beads of different sizes in roughly equimolar concentrations and is designed to calibrate the size of particles measured by macro mass photometry, including adenoviral and lentiviral vectors. Each package contains a 0.3 mL vial of silica beads suspended in deionized water.
Catalog No. NC3737117 Supplier REFEYN INC Supplier No. MPCON41030
SizeFerence is a calibrant tailored for the KaritroMP. This contains a mix of silica beads of different sizes in roughly equimolar concentrations and is designed to calibrate the size of particles measured by macro mass photometry, including adenoviral and lentiviral vectors. Each package contains a 0.3 mL vial of silica beads suspended in deionized water.