Learn More
Mti Corporation SIZERO24D10C1-CAVITY-10D
SDP
Catalog No. NC1516944
Encompass
Zero Diffraction Plate for XRD: 24.6 Dia x1.0 t mm w/ Cavity 10 ID x0.2 mm, Si Crystal - SiZero24D10C1-cavity-10D
Catalog No. NC1516944 Supplier Mti Corporation Supplier No. ZEROSI24D10C1CAVITY10D
Zero Diffraction Plate for XRD: 24.6 Dia x1.0 t mm w/ Cavity 10 ID x0.2 mm, Si Crystal - SiZero24D10C1-cavity-10D