Learn More
OHLHEISER Jun-Air SJ-27F oil, 16oz Bottle
Catalog No. NC0342985
Encompass
This item has a minimum order quantity of 4 per supplier requirements. Jun-Air SJ-27F oil. Specially formulated oil used in all lubricated Jun-Air compressors. Prevents corrosion, assists in cooling and extends valve life. Jun-Air recommends that the oil be changed every two years at a minimum. Used on M3, M4, M6, M12, M18, M24, M36 models. 16oz bottle
Catalog No. NC0342985 Supplier OHLHEISER Supplier No. 5631000
This item has a minimum order quantity of 4 per supplier requirements. Jun-Air SJ-27F oil. Specially formulated oil used in all lubricated Jun-Air compressors. Prevents corrosion, assists in cooling and extends valve life. Jun-Air recommends that the oil be changed every two years at a minimum. Used on M3, M4, M6, M12, M18, M24, M36 models. 16oz bottle