Learn More
BAY ADVANCED TECHNOLOGIES LLC SJ-27F OIL, 16 oz
Supplier:  BAY ADVANCED TECHNOLOGIES LLC 5631000
Encompass
Jun-Air SJ-27F oil. Specially formulated oil used in all lubricated Jun-Air compressors. Prevents corrosion, assists in cooling and extends valve life.
Catalog No. NC9057134