Learn More
Cayman Chemical SJ000291942 10mg
SDP
Supplier:  Cayman Chemical 2990410 MG
Encompass_Preferred
A BMP signaling activator; activates BMP4 in C33A-2D2 human cervical carcinoma clonal reporter cells (EC50 = ≤1 µM); induces ventralization, as well as increases expression of the BMP target genes bmp2b and szl and the cooperatively-induced BMP target genes vent and vox in zebrafish embryos; induces BMP-dependent osteoblastic differentiation of C2C12 murine myoblasts
Catalog No. 50-298-8941