Learn More
Medchemexpress LLC HY-112331 5mg Medchemexpress, SJ000291942 CAS:425613-09-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1123315MG
Encompass_Preferred
Medchemexpress, HY-112331 5mg SJ000291942 CAS:425613-09-8 SJ000291942 is an activator of the canonical bone morphogenetic proteins (BMP) signaling pathway. BMPs are members of the transforming growth factor beta (TGFβ) family of secreted signaling molecules. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9083