Learn More
Medchemexpress LLC SJ1008030 TFA | 2863634-97-1 | 99.8% | 987.96 | 1 MG
SDP
Catalog No. 5000361419
Encompass_Preferred
SJ1008030 TFA is a JAK2 PROTAC designed for selective degradation of JAK2. This compound inhibits MHH-CALL-4 cell growth and is suitable for leukemia research. It degrades JAKs, GSPT1, and IKZF1 in a dose-dependent manner, indicating inhibition of the JAK-STAT signaling pathway.
  • Selectively degrades JAK2
  • Inhibits MHH-CALL-4 cell growth with an IC50 of 5.4 nM
  • Can be used for leukemia research
  • Degrades JAKs, GSPT1, and IKZF1 in a dose-dependent manner
  • Indicates inhibition of the JAK-STAT signaling pathway
Catalog No. 50-003-61419 Supplier Medchemexpress LLC Supplier No. HY147330A1MG
SJ1008030 TFA is a JAK2 PROTAC designed for selective degradation of JAK2. This compound inhibits MHH-CALL-4 cell growth and is suitable for leukemia research. It degrades JAKs, GSPT1, and IKZF1 in a dose-dependent manner, indicating inhibition of the JAK-STAT signaling pathway.
  • Selectively degrades JAK2
  • Inhibits MHH-CALL-4 cell growth with an IC50 of 5.4 nM
  • Can be used for leukemia research
  • Degrades JAKs, GSPT1, and IKZF1 in a dose-dependent manner
  • Indicates inhibition of the JAK-STAT signaling pathway