Learn More
Medchemexpress LLC SJ1008030 TFA | 2863634-97-1 | 99.81% | 987.96 | 25 MG
SDP
Catalog No. 5000362604
Encompass_Preferred
SJ1008030 TFA is a JAK2 PROTAC designed to selectively degrade JAK2. This compound has demonstrated potent inhibitory effects on MHH-CALL-4 cell growth with an IC50 of 5.4 nM, making it a valuable tool for leukemia research.
  • Selectively degrades JAK2.
  • Inhibits MHH-CALL-4 cell growth with an IC50 of 5.4 nM.
  • Useful for leukemia research.
  • Degrades JAKs, GSPT1, and IKZF1 in a dose-dependent manner in MHH-CALL-4 cells.
  • Dose-dependently degrades JAK2 and GSPT1 protein in xenograft bone marrow SJBALL021415 cells, indicating the inhibition of the JAK-STAT signaling pathway.
Catalog No. 50-003-62604 Supplier Medchemexpress LLC Supplier No. 25MGHY147330A
SJ1008030 TFA is a JAK2 PROTAC designed to selectively degrade JAK2. This compound has demonstrated potent inhibitory effects on MHH-CALL-4 cell growth with an IC50 of 5.4 nM, making it a valuable tool for leukemia research.
  • Selectively degrades JAK2.
  • Inhibits MHH-CALL-4 cell growth with an IC50 of 5.4 nM.
  • Useful for leukemia research.
  • Degrades JAKs, GSPT1, and IKZF1 in a dose-dependent manner in MHH-CALL-4 cells.
  • Dose-dependently degrades JAK2 and GSPT1 protein in xenograft bone marrow SJBALL021415 cells, indicating the inhibition of the JAK-STAT signaling pathway.