Learn More
TARGETMOL CHEMICALS INC SJ572403 5MG
SDP
Catalog No. 502581140
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SJ572403 is an inhibitor of disordered protein p27 with Kd of 2.2 mM for specific regions within the D2 subdomain of p27-KID. SJ572403 can be used in studies about diseases with intrinsically disordered proteins. purity: 99%
Catalog No. 50-258-1140 Supplier TARGETMOL CHEMICALS INC Supplier No. T168895MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SJ572403 is an inhibitor of disordered protein p27 with Kd of 2.2 mM for specific regions within the D2 subdomain of p27-KID. SJ572403 can be used in studies about diseases with intrinsically disordered proteins. purity: 99%