Learn More
TARGETMOL CHEMICALS INC SJ6986 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4125725MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SJ6986 is a potent selective and orally active GSPT1/2 degrader. SJ6986 degrades GSPT1 with a DC 50 of 2.1 nM (D max 99%) [1]. purity: 99%
Catalog No. 50-257-9944