Learn More
Apexbio Technology LLC SJB2-043 63388-44-3 5mg
SDP
Catalog No. 502240451
Encompass_Preferred
SJB2-043 (CAS 63388-44-3) is a small molecule inhibitor of ubiquitin-specific protease 1 (USP1) a deubiquitinating enzyme involved in the regulation of DNA repair by deubiquitinating key proteins in the Fanconi anemia (FA) pathway SJB2-043 exhibits an IC50 of 0 544 M for USP1 In cellular models such as K562 leukemia cells and primary AML cells SJB2-043 reduces USP1 levels and promotes proteasomal degradation of ID1 as well as decreasing ID2 and ID3 protein levels Treatment with SJB2-043 also increases Ub-FANCD2 and Ub-PCNA in HeLa cells reflecting modulation of DNA repair processes This compound serves as a valuable tool for studies investigating USP1 function and the regulation of ID proteins in cancer and stem cell biology
This item is not returnable. View return policy
Catalog No. 50-224-0451 Supplier Apexbio Technology LLC Supplier No. A38235
SJB2-043 (CAS 63388-44-3) is a small molecule inhibitor of ubiquitin-specific protease 1 (USP1) a deubiquitinating enzyme involved in the regulation of DNA repair by deubiquitinating key proteins in the Fanconi anemia (FA) pathway SJB2-043 exhibits an IC50 of 0 544 M for USP1 In cellular models such as K562 leukemia cells and primary AML cells SJB2-043 reduces USP1 levels and promotes proteasomal degradation of ID1 as well as decreasing ID2 and ID3 protein levels Treatment with SJB2-043 also increases Ub-FANCD2 and Ub-PCNA in HeLa cells reflecting modulation of DNA repair processes This compound serves as a valuable tool for studies investigating USP1 function and the regulation of ID proteins in cancer and stem cell biology