Learn More
Medchemexpress LLC HY-80012 5mg Medchemexpress, SJB3-019A CAS:2070015-29-9 Purity:>98%
SDP
Catalog No. 502030993
Encompass_Preferred
Medchemexpress, HY-80012 5mg SJB3-019A CAS:2070015-29-9 SJB3-019A is a potent and novel USP1 inhibitor, 5 times more potent than SJB2-043 in promoting ID1 degradation and cytoxicity in K562 cells with IC50 of 0.0781 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0993 Supplier Medchemexpress LLC Supplier No. HY800125MG
Medchemexpress, HY-80012 5mg SJB3-019A CAS:2070015-29-9 SJB3-019A is a potent and novel USP1 inhibitor, 5 times more potent than SJB2-043 in promoting ID1 degradation and cytoxicity in K562 cells with IC50 of 0.0781 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.