Learn More
TARGETMOL CHEMICALS INC SJB3-019A 5MG
SDP
Catalog No. 502581396
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SJB3-019A is a potent potent and novel ubiquitin-specific protease 1 (USP1) inhibitor which promotes ID1 degradation and cytotoxicity in K562 cells 5 times more than SJB2-043 with IC50 of 0.0781 uM. SJB3-019A inhibits cell proliferation and causes cell apoptosis. purity: 99%
Catalog No. 50-258-1396 Supplier TARGETMOL CHEMICALS INC Supplier No. T129265MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SJB3-019A is a potent potent and novel ubiquitin-specific protease 1 (USP1) inhibitor which promotes ID1 degradation and cytotoxicity in K562 cells 5 times more than SJB2-043 with IC50 of 0.0781 uM. SJB3-019A inhibits cell proliferation and causes cell apoptosis. purity: 99%