Learn More
ATCC SJCRH30 (RMS 13); Rhabdomyosarcoma; Human (Homo sapiens)
Catalog No. 502384697
Encompass
SJCRH30 [RC13, RMS 13, SJRH30] is a cell line exhibiting fibroblast-like morphology that was isolated from the muscle of a 17-year-old, White male patient with rhabdomyosarcoma. This cell line was deposited by DN Shapiroand and can be used in immunology research.
Catalog No. 50-238-4697 Supplier ATCC Supplier No. CRL2061
SJCRH30 [RC13, RMS 13, SJRH30] is a cell line exhibiting fibroblast-like morphology that was isolated from the muscle of a 17-year-old, White male patient with rhabdomyosarcoma. This cell line was deposited by DN Shapiroand and can be used in immunology research.