Learn More
Medchemexpress LLC Sjf-1521 | 2230821-40-4 | >98.0% | C57H61ClFN7O9S | 10MG
SDP
Catalog No. 5000260837
Encompass_Preferred
SJF-1521 is a selective EGFR PROTAC degrader that induces proteasomal degradation of EGFR in OVCAR8 cells and is used for tumor research and cellular pathway studies. It couples an EGFR-binding ligand to a VHL-recruiting moiety to promote targeted ubiquitination and clearance of the receptor.
  • Selective EGFR degrader enabling modulation of receptor levels.
  • Demonstrated to induce EGFR degradation in OVCAR8 cells.
  • Dual mechanism: EGFR binding and VHL recruitment for proteasomal degradation.
  • Suitable for tumor biology and signal transduction research in cell models.
  • Reported high purity suitable for biochemical and cellular assays (≥98% HPLC).
  • Available in multiple research-scale pack sizes for experimental flexibility.
Catalog No. 50-002-60837 Supplier Medchemexpress LLC Supplier No. HY13186510MG
Add to Cart
Add to Cart
SJF-1521 is a selective EGFR PROTAC degrader that induces proteasomal degradation of EGFR in OVCAR8 cells and is used for tumor research and cellular pathway studies. It couples an EGFR-binding ligand to a VHL-recruiting moiety to promote targeted ubiquitination and clearance of the receptor.
  • Selective EGFR degrader enabling modulation of receptor levels.
  • Demonstrated to induce EGFR degradation in OVCAR8 cells.
  • Dual mechanism: EGFR binding and VHL recruitment for proteasomal degradation.
  • Suitable for tumor biology and signal transduction research in cell models.
  • Reported high purity suitable for biochemical and cellular assays (≥98% HPLC).
  • Available in multiple research-scale pack sizes for experimental flexibility.