Learn More
Medchemexpress LLC HY-133137 5mg , SJF620 CAS:2376187-16-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1331375MG
Encompass_Preferred
HY-133137 5mg SJF620 CAS: 2376187-16-3 SJF620 is a PROTAC connected by ligands for Cereblon and Btk with a DC50 of 7.9 nM. SJF620 contains a Lenalidomide analog for recruiting CRBN. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9492