Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SJG-136 | 232931-57-6 | 99.5% | 556.61 | C31H32N4O6 | 50 MG
SDP

Catalog No. 5000204308
Encompass_Preferred

SJG-136 is a pyrrolobenzodiazepine-class DNA cross-linking agent supplied as a research reagent. It exhibits potent antitumor activity and an XL50 of 45 nM against pBR322 DNA. The compound is a solid (white to yellow), with formula C31H32N4O6, molecular weight 556.61, and reported purity of 99.47%.

  • Pyrrolobenzodiazepine DNA cross-linker with potent antitumor activity.
  • XL50 for pBR322 DNA: 45 nM.
  • CAS number 232931-57-6.
  • Chemical formula C31H32N4O6; molecular weight 556.61.
  • Purity reported as 99.47% (certificate of analysis available).
  • Appearance: solid, white to yellow.
  • Available in multiple package sizes including 50 mg and solution formats.
  • Associated documentation available: datasheet, SDS, COA, HNMR, LCMS.

Catalog No. 50-002-04308 Supplier Medchemexpress LLC Supplier No. HY1457350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SJG-136 is a pyrrolobenzodiazepine-class DNA cross-linking agent supplied as a research reagent. It exhibits potent antitumor activity and an XL50 of 45 nM against pBR322 DNA. The compound is a solid (white to yellow), with formula C31H32N4O6, molecular weight 556.61, and reported purity of 99.47%.

  • Pyrrolobenzodiazepine DNA cross-linker with potent antitumor activity.
  • XL50 for pBR322 DNA: 45 nM.
  • CAS number 232931-57-6.
  • Chemical formula C31H32N4O6; molecular weight 556.61.
  • Purity reported as 99.47% (certificate of analysis available).
  • Appearance: solid, white to yellow.
  • Available in multiple package sizes including 50 mg and solution formats.
  • Associated documentation available: datasheet, SDS, COA, HNMR, LCMS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.