Learn More
Electron Microscopy Sciences Veco Sjostrand Grid Serial Section R100/200A, Copper
SDP
Catalog No. 5028670
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Pitch: 250 µm
Hole: 120 µm
Bar: 130 µm
Catalog No. 50-286-70 Supplier Electron Microscopy Sciences Supplier No. R12CACU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Pitch: 250 µm
Hole: 120 µm
Bar: 130 µm