Learn More
Electron Microscopy Sciences Veco Sjostrand Grid Serial Section R100/200A, Ni
SDP
Catalog No. 5028671
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel
Pitch: 250 µm
Hole: 75 µm
Bar: 50 µm
Catalog No. 50-286-71 Supplier Electron Microscopy Sciences Supplier No. R12CANI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Nickel
Pitch: 250 µm
Hole: 75 µm
Bar: 50 µm