Learn More
Trajan Scientific SK-10F-C/T-5/0.63C SYR 6PK
SDP
Catalog No. 502530366
Encompass_Preferred
Trajan PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Diameter Outer Needle: 0.63 mm, 0.024 in., Needle Gauge: 23 G, Needle Point Style: Cone, Length Needle: 50 mm, 1.96 in., Volume: 10 uL, Connection Type: Fixed, Description: Not gas tight, Diameter Inner Needle: 0.004 in., 0.11 mm
Catalog No. 50-253-0366 Supplier Trajan Scientific Supplier No. 002971
Trajan PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Diameter Outer Needle: 0.63 mm, 0.024 in., Needle Gauge: 23 G, Needle Point Style: Cone, Length Needle: 50 mm, 1.96 in., Volume: 10 uL, Connection Type: Fixed, Description: Not gas tight, Diameter Inner Needle: 0.004 in., 0.11 mm