Learn More
Trajan Scientific SK-10F-C/T-GT-5/0.63C SYR 6PK
SDP
Supplier:  Trajan Scientific 002972
Encompass_Preferred
SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Description: Gas tight, Volume: 10 uL, Diameter Outer Needle: 0.63 mm, 0.024 in., Needle Gauge: 23 G, Length Needle: 50 mm, 1.96 in., Connection Type: Fixed, Diameter Inner Needle: 0.004 in., 0.11 mm
Catalog No. 50-253-0365