Learn More
Medchemexpress LLC SK-575 | 2523016-96-6 | 99.9% | 876.97 | 25 MG
SDP
Catalog No. 5000349888
Encompass_Preferred
SK-575 is a highly potent and specific proteolysis-targeting chimera (PROTAC) degrader of PARP1, with an IC50 of 2.30 nM. It potently inhibits the growth of cancer cells bearing BRCA1/2 mutations.
  • Potently inhibits the growth of cancer cells bearing BRCA1/2 mutations.
  • Shows good PARP1 degradation activity in cancer cell lines.
  • Effectively induces the formation of γH2AX in MDA-MB-436 and Capan-1 cells in a dose-dependent manner.
  • Significantly inhibits tumor growth in HR-deficient xenograft models.
  • Achieves sufficient plasma exposure and effectively induces PARP1 degradation in tumor tissue.
  • Well tolerated in animal models.
Catalog No. 50-003-49888 Supplier Medchemexpress LLC Supplier No. 25MGHY139156
SK-575 is a highly potent and specific proteolysis-targeting chimera (PROTAC) degrader of PARP1, with an IC50 of 2.30 nM. It potently inhibits the growth of cancer cells bearing BRCA1/2 mutations.
  • Potently inhibits the growth of cancer cells bearing BRCA1/2 mutations.
  • Shows good PARP1 degradation activity in cancer cell lines.
  • Effectively induces the formation of γH2AX in MDA-MB-436 and Capan-1 cells in a dose-dependent manner.
  • Significantly inhibits tumor growth in HR-deficient xenograft models.
  • Achieves sufficient plasma exposure and effectively induces PARP1 degradation in tumor tissue.
  • Well tolerated in animal models.