Learn More
Promocell GmbH PromoCell Skeletal Muscle Cell Differentiation Medium 500 ml, ready-to-use, serum-free, with C-39366 attached to the flask
Catalog No. 50306216
Encompass_Preferred
The Skeletal Muscle Cell Differentiation Medium is a defined and animal-component free medium developed to induce the fusion of skeletal muscle cells to myotubes with typical multinucleated syncytia.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine, rat, canine, and rabbit skeletal muscle cells.| Media supplement | Final supplement concentration || Insulin (recombinant human) | 10 μg / ml |
Catalog No. 50-306-216 Supplier Promocell GmbH Supplier No. C23061
The Skeletal Muscle Cell Differentiation Medium is a defined and animal-component free medium developed to induce the fusion of skeletal muscle cells to myotubes with typical multinucleated syncytia.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine, rat, canine, and rabbit skeletal muscle cells.| Media supplement | Final supplement concentration || Insulin (recombinant human) | 10 μg / ml |