Learn More
Promocell GmbH PromoCell Skeletal Muscle Cell Growth Medium 500 ml, ready-to-use, low serum, with C-39365
Catalog No. 50306215
Encompass_Preferred
The PromoCell Skeletal Muscle Cell Growth Medium is a low-serum medium (5% V/V) developed for the expansion of human skeletal muscle cell cultures.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine, rat, canine, and rabbit skeletal muscle cells.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.05 ml / ml || Fetuin (bovine) | 50 μg / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 10 μg / ml || Dexamethasone | 0.4 μg / ml |
Catalog No. 50-306-215 Supplier Promocell GmbH Supplier No. C23060
The PromoCell Skeletal Muscle Cell Growth Medium is a low-serum medium (5% V/V) developed for the expansion of human skeletal muscle cell cultures.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine, rat, canine, and rabbit skeletal muscle cells.| Media supplement | Final supplement concentration || Fetal Calf Serum | 0.05 ml / ml || Fetuin (bovine) | 50 μg / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 10 μg / ml || Dexamethasone | 0.4 μg / ml |