Learn More
Medchemexpress LLC SK1-I (BML-258) | 1072443-89-0 | 98.8% | 277.40 g·mol⁻¹ | C17H27NO2 | 50 MG
SDP
Catalog No. 5000228334
Encompass_Preferred
SK1-I (BML-258) is a sphingosine analog and an isozyme-selective, competitive inhibitor of sphingosine kinase 1 (SPHK1). It has a reported Ki of approximately 10 μM and has been shown to enhance autophagy and exhibit antitumor activity in preclinical studies. Supplied as a 50 mg powder for research use only.
  • Isozyme-selective competitive SPHK1 inhibitor (Ki ≈ 10 μM)
  • Sphingosine analog with autophagy-enhancing effects
  • Reported antitumor activity in preclinical models
  • High purity (98.83%) suitable for research applications
  • Storage: powder stable at -20 °C (3 years) or 4 °C (2 years)
Catalog No. 50-002-28334 Supplier Medchemexpress LLC Supplier No. HY11901650MG
SK1-I (BML-258) is a sphingosine analog and an isozyme-selective, competitive inhibitor of sphingosine kinase 1 (SPHK1). It has a reported Ki of approximately 10 μM and has been shown to enhance autophagy and exhibit antitumor activity in preclinical studies. Supplied as a 50 mg powder for research use only.
  • Isozyme-selective competitive SPHK1 inhibitor (Ki ≈ 10 μM)
  • Sphingosine analog with autophagy-enhancing effects
  • Reported antitumor activity in preclinical models
  • High purity (98.83%) suitable for research applications
  • Storage: powder stable at -20 °C (3 years) or 4 °C (2 years)