Learn More
ECM Bioscience Sphingosine Kinase 2 Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience SK6590
Encompass_Preferred
The Sphingosine Kinase 2 (SK2) phospho-regulation antibody kit can be used to detect phosphorylation of Thr-578 relative to total SK2 expression. The kit includes rabbit polyclonal antibodies, blocking peptides, and secondary reagents for detection in western blot. Applications: WB, E; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8839